Thermo Scientific Chemicals

Iron(II) sulfate heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A15178.0E
2500 g, Each
Thermo Scientific Chemicals

Iron(II) sulfate heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A15178.0E
2500 g, Each
Quantity
Catalog number: A15178.0E
also known as A15178-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-63-0
IUPAC Name
λ²-iron(2+) heptahydrate sulfate
Molecular Formula
FeH14O11S
InChI Key
SURQXAFEQWPFPV-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
Appearance (Color)
Pale green to blue-green
Form
Crystals or crystalline powder
Assay (Redox Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
40-49%

Description

Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Air and moisture sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text