Thermo Scientific Chemicals

(1-Hexadecyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A15180.06
5 g, Each
Thermo Scientific Chemicals

(1-Hexadecyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A15180.06
5 g, Each
Quantity
Catalog number: A15180.06
also known as A15180-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14866-43-4
IUPAC Name
hexadecyltriphenylphosphanium bromide
Molecular Formula
C34H48BrP
InChI Key
UXMZNEHSMYESLH-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Water Content (Karl Fischer Titration)
≤1%
Appearance (Color)
White
Form
Powder or lumps
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%
Melting Point (clear melt)
95.0-102.0?C

Description

Widely used as preservative, water treatment in agriculture. Also used as antistatic agent in textiles, fibers, leather, as flocculants, deodorant and as phase transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used as preservative, water treatment in agriculture. Also used as antistatic agent in textiles, fibers, leather, as flocculants, deodorant and as phase transfer catalyst.

Solubility
Soluble in ethanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text