Thermo Scientific Chemicals

Tetrahydrofurfuryl bromide, 95%, Thermo Scientific Chemicals

Catalog number: A15192.09
10 g, Each
Thermo Scientific Chemicals

Tetrahydrofurfuryl bromide, 95%, Thermo Scientific Chemicals

Catalog number: A15192.09
10 g, Each
Quantity
Catalog number: A15192.09
also known as A15192-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1192-30-9
IUPAC Name
2-(bromomethyl)oxolane
Molecular Formula
C5H9BrO
InChI Key
VOHILFSOWRNVJJ-UHFFFAOYNA-N
SMILES
BrCC1CCCO1
Assay (GC)
≥94.0%
Form
Liquid
Refractive Index
1.4840-1.4900 @ 20?C
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text