Tetrahydrofurfuryl bromide, 95%, Thermo Scientific Chemicals
Tetrahydrofurfuryl bromide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofurfuryl bromide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A15192.18
also known as A15192-18
Price (USD)/ Each
192.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
192.00
Add to cart
Tetrahydrofurfuryl bromide, 95%, Thermo Scientific Chemicals
Catalog numberA15192.18
Price (USD)/ Each
192.00
-
Add to cart
Chemical Identifiers
CAS1192-30-9
IUPAC Name2-(bromomethyl)oxolane
Molecular FormulaC5H9BrO
InChI KeyVOHILFSOWRNVJJ-UHFFFAOYNA-N
SMILESBrCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
FormLiquid
Refractive Index1.4840-1.4900 @ 20?C
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only