3-Bromo-4-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals
3-Bromo-4-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15198.06
also known as A15198-06
Price (USD)/ Each
46.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
46.70
Add to cart
3-Bromo-4-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberA15198.06
Price (USD)/ Each
46.70
-
Add to cart
Chemical Identifiers
CAS77771-02-9
IUPAC Name3-bromo-4-fluorobenzaldehyde
Molecular FormulaC7H4BrFO
InChI KeyFAHZIKXYYRGSHF-UHFFFAOYSA-N
SMILESFC1=CC=C(C=O)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale yellow
FormFused solid or clear liquid as melt
Free acid (titration)≤1%
Melting Point (clear melt)28.0-34.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only