Thermo Scientific Chemicals

2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: A15211.18
50 g, Each
Thermo Scientific Chemicals

2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: A15211.18
50 g, Each
Quantity
Catalog number: A15211.18
also known as A15211-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-25-4
IUPAC Name
2-(2-methoxyphenyl)acetic acid
Molecular Formula
C9H10O3
InChI Key
IVEWTCACRDEAOB-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1CC(O)=O
Assay (HPLC)
≥98.5%
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5% to ≤101.5%
Identification (FTIR)
Conforms

Description

2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water. Also soluble in methanol, ether, ethanol and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text