2,4-Difluorobenzyl bromide, 98+%, Thermo Scientific Chemicals
2,4-Difluorobenzyl bromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Difluorobenzyl bromide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A15213.14
also known as A15213-14
Price (USD)/ Each
206.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
206.00
Add to cart
2,4-Difluorobenzyl bromide, 98+%, Thermo Scientific Chemicals
Catalog numberA15213.14
Price (USD)/ Each
206.00
-
Add to cart
Chemical Identifiers
CAS23915-07-3
IUPAC Name1-(bromomethyl)-2,4-difluorobenzene
Molecular FormulaC7H5BrF2
InChI KeyIBLMYGXJKQIGSN-UHFFFAOYSA-N
SMILESFC1=CC=C(CBr)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)>98.0%
Refractive Index1.5225-1.5275 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only