Thermo Scientific Chemicals

Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A15215.14
25 g, Each
Thermo Scientific Chemicals

Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A15215.14
25 g, Each
Quantity
Catalog number: A15215.14
also known as A15215-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-34-8
IUPAC Name
tetraethylazanium chloride
Molecular Formula
C8H20ClN
InChI Key
YMBCJWGVCUEGHA-UHFFFAOYSA-M
SMILES
[Cl-].CC[N+](CC)(CC)CC
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (Argentometric Titration)
≥97.5 to ≤102.5% (on dried substance)
Appearance (Color)
White
Loss on Drying
≤10%

Description

Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (1410 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text