Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals
Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A15215.22
also known as A15215-22
Price (USD)/ Each
72.65
Online exclusive
81.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
81.10 
Save 8.45 (10%)
Add to cart
Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals
Catalog numberA15215.22
Price (USD)/ Each
72.65
Online exclusive
81.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
CAS56-34-8
IUPAC Nametetraethylazanium chloride
Molecular FormulaC8H20ClN
InChI KeyYMBCJWGVCUEGHA-UHFFFAOYSA-M
SMILES[Cl-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (Argentometric Titration)≥97.5 to ≤102.5% (on dried substance)
Appearance (Color)White
Loss on Drying≤10%
Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (1410 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Kenneth M.Harmon; Julie M.Gabriele; Jennifer Harmon. Hydrogen bonding Part 30. New IR spectra-structure correlations for tetraethylammonium, tetramethylammonium, and N,N-dimethyl-pyrrolidinium fluoride monohydrates, tetraethylammonium chloride monohydrate, and tetramethylammonium hydroxide dihydrate; evidence for a planar (H2O·F-)2 cluster. Journal of Molecular Structure. 1990, 53-62.
  2. Dimitra Kovala-Demertzi; Nikolaos Kourkoumelis; Kinga Derlat; Julita Michalak; Fotini J.Andreadaki; Ioannis D.Kostas. title. Inorganica Chimica Acta. 2008, 361,(5), 1562-1565.