Thermo Scientific Chemicals

Methyl 3,5-dihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A15224.30
250 g, Each
Thermo Scientific Chemicals

Methyl 3,5-dihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A15224.30
250 g, Each
Quantity
Catalog number: A15224.30
also known as A15224-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2150-44-9
IUPAC Name
methyl 3,5-dihydroxybenzoate
Molecular Formula
C8H8O4
InChI Key
RNVFYQUEEMZKLR-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(O)=CC(O)=C1
Appearance (Color)
White to cream or pale brown
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
164.0-170.0?C

Description

Methyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text