Thermo Scientific Chemicals

Benzo[b]furan, 97+%, Thermo Scientific Chemicals

Catalog number: A15225.09
10 g, Each
Thermo Scientific Chemicals

Benzo[b]furan, 97+%, Thermo Scientific Chemicals

Catalog number: A15225.09
10 g, Each
Quantity
Catalog number: A15225.09
also known as A15225-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
271-89-6
IUPAC Name
1-benzofuran
Molecular Formula
C8H6O
InChI Key
IANQTJSKSUMEQM-UHFFFAOYSA-N
SMILES
O1C=CC2=CC=CC=C12
Assay (GC)
≥97.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.5635-1.5685 @ 20?C
Form
Liquid

Description

Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text