Thermo Scientific Chemicals

1,3-Diphenylguanidine, 97%, Thermo Scientific Chemicals

Catalog number: A15234.0B
1000 g, Each
Thermo Scientific Chemicals

1,3-Diphenylguanidine, 97%, Thermo Scientific Chemicals

Catalog number: A15234.0B
1000 g, Each
Quantity
Catalog number: A15234.0B
also known as A15234-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-06-7
IUPAC Name
N,N''-diphenylguanidine
Molecular Formula
C13H13N3
InChI Key
OWRCNXZUPFZXOS-UHFFFAOYSA-N
SMILES
NC(NC1=CC=CC=C1)=NC1=CC=CC=C1
Appearance (Color)
White to pale cream to pale brown
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
142.0-152.0?C

Description

1,3-Diphenylguanidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylguanidine is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text