1,3-Diphenylguanidine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Diphenylguanidine is used as pharmaceutical intermediate.
Solubility
Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- P.V.Shah; M.R.Sumler; Y.M.Ioannou; H.L.Fisher; L.L.Hall. Dermal absorption and disposition of 1,3-diphenylguanidine in rats. Journal of Toxicology and Environmental Health. 1985, 15,(5), 623-633.
- P.V.Gushchin; M.Haukka; N.A.Bokach; V.Yu.Kukushkin. Platinum(iv)-mediated nucleophilic addition of 1,3-diphenylguanidine to propiononitrile. Russian Chemical Bulletin. 2008, 57,(10), 2125-2131.