Thermo Scientific Chemicals

4-Nitrobenzyl bromide, 97+%, Thermo Scientific Chemicals

Catalog number: A15236.14
25 g, Each
Thermo Scientific Chemicals

4-Nitrobenzyl bromide, 97+%, Thermo Scientific Chemicals

Catalog number: A15236.14
25 g, Each
Quantity
Catalog number: A15236.14
also known as A15236-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-11-8
IUPAC Name
1-(bromomethyl)-4-nitrobenzene
Molecular Formula
C7H6BrNO2
InChI Key
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(CBr)C=C1
Appearance (Color)
White to cream or pale yellow
Form
Crystalline powder
Assay (GC)
>97.0%
Solution Test
5% in acetone: clear - no visible particlulates

Description

4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

Solubility
It hydrolyzes in water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text