Thermo Scientific Chemicals

4-Acetylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15239.06
5 g, Each
Thermo Scientific Chemicals

4-Acetylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15239.06
5 g, Each
Quantity
Catalog number: A15239.06
also known as A15239-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-89-0
IUPAC Name
4-acetylbenzoic acid
Molecular Formula
C9H8O3
InChI Key
QBHDSQZASIBAAI-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(C=C1)C(O)=O
Assay (HPLC)
≥98.0%
Appearance (Color)
White to pale cream
Form
Powder
Melting Point (clear melt)
209.0-216.0?C
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text