Thermo Scientific Chemicals

2-Phenylethanol, 98+%, Thermo Scientific Chemicals

Catalog number: A15241.0I
5000 g, Each
Thermo Scientific Chemicals

2-Phenylethanol, 98+%, Thermo Scientific Chemicals

Catalog number: A15241.0I
5000 g, Each
Quantity
Catalog number: A15241.0I
also known as A15241-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-12-8
IUPAC Name
2-phenylethan-1-ol
Molecular Formula
C8H10O
InChI Key
WRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILES
OCCC1=CC=CC=C1
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.5310-1.5350 @ 20?C
Appearance (Color)
Clear colorless

Description

2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.

Solubility
Miscible with chloroform.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text