Thermo Scientific Chemicals

6-Methyl-1,2,3,4-tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A15245.09
10 g, Each
Thermo Scientific Chemicals

6-Methyl-1,2,3,4-tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A15245.09
10 g, Each
Quantity
Catalog number: A15245.09
also known as A15245-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-61-2
Appearance (Color)
White to yellow to pale brown or clear colorless to yellow to pale brown
Form
Fused solid or liquid as melt
Assay (GC)
≥97.5%
Melting Point (clear melt)
32.0-38.0?C

Description

6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text