Thermo Scientific Chemicals

Triisopropyl phosphite, 90+%, Thermo Scientific Chemicals

Catalog number: A15247.AP
500 mL, Each
Thermo Scientific Chemicals

Triisopropyl phosphite, 90+%, Thermo Scientific Chemicals

Catalog number: A15247.AP
500 mL, Each
Quantity
Catalog number: A15247.AP
also known as A15247-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
116-17-6
IUPAC Name
tris(propan-2-yl) phosphite
Molecular Formula
C9H21O3P
InChI Key
SJHCUXCOGGKFAI-UHFFFAOYSA-N
SMILES
CC(C)OP(OC(C)C)OC(C)C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥90.0%
Identification (FTIR)
Conforms
Refractive Index
1.4075-1.4145 @20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text