Thermo Scientific Chemicals

Indole-3-butyric acid, 98%, Thermo Scientific Chemicals

Catalog number: A15260.14
25 g, Each
Thermo Scientific Chemicals

Indole-3-butyric acid, 98%, Thermo Scientific Chemicals

Catalog number: A15260.14
25 g, Each
Quantity
Catalog number: A15260.14
also known as A15260-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
133-32-4
IUPAC Name
4-(1H-indol-3-yl)butanoic acid
Molecular Formula
C12H13NO2
InChI Key
JTEDVYBZBROSJT-UHFFFAOYSA-N
SMILES
OC(=O)CCCC1=CNC2=CC=CC=C12
Appearance (Color)
White to cream to yellow to pale brown to brown or pink
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

Solubility
Soluble in water(0.25g/L).

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text