3,4-Dimethoxybenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
3,4-Dimethoxybenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dimethoxybenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15263.06
also known as A15263-06
Price (USD)/ Each
71.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
71.50
Add to cart
3,4-Dimethoxybenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA15263.06
Price (USD)/ Each
71.50
-
Add to cart
Chemical Identifiers
CAS23095-31-0
IUPAC Name3,4-dimethoxybenzene-1-sulfonyl chloride
Molecular FormulaC8H9ClO4S
InChI KeyRSJSYCZYQNJQPY-UHFFFAOYSA-N
SMILESCOC1=C(OC)C=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream to pale grey
FormPowder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only