ADA, 98+%
Thermo Scientific Chemicals

ADA, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A15267.22
also known as A15267-22
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
Add to cart
ADA, 98+%
Catalog numberA15267.22
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS26239-55-4
IUPAC Name2-[(carbamoylmethyl)(carboxylatomethyl)azaniumyl]acetate
Molecular FormulaC6H9N2O5
InChI KeyQZTKDVCDBIDYMD-UHFFFAOYSA-M
SMILESNC(=O)C[NH+](CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. P.G.Righetti; M.Chiari; C.Gelfi. Immobilized pH gradients: effect of salts, added carrier ampholytes and voltage gradients on protein patterns. Electrophoresis. 1988, 9,(2), 65-73.
  2. Ting-Chien Chen; Andrew Hong. Chelating extraction of lead and copper from an authentic contaminated soil using N-(2-acetamido)iminodiacetic acid and S-carboxymethyl-l-cysteine. Journal of Hazardous Materials. 1995, 41,(2-3), 147-160.
  3. Biological buffer, pKa = 6.9 at 20°: Biochemistry, 5, 467 (1966).