Thermo Scientific Chemicals

ADA, 98+%

Catalog number: A15267.36
500 g, Each
Thermo Scientific Chemicals

ADA, 98+%

Catalog number: A15267.36
500 g, Each
Quantity
Catalog number: A15267.36
also known as A15267-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26239-55-4
IUPAC Name
2-[(carbamoylmethyl)(carboxylatomethyl)azaniumyl]acetate
Molecular Formula
C6H9N2O5
InChI Key
QZTKDVCDBIDYMD-UHFFFAOYSA-M
SMILES
NC(=O)C[NH+](CC([O-])=O)CC([O-])=O
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text