Thermo Scientific Chemicals

4-(Trifluoromethyl)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15276.14
25 g, Each
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15276.14
25 g, Each
Quantity
Catalog number: A15276.14
also known as A15276-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
455-19-6
IUPAC Name
4-(trifluoromethyl)benzaldehyde
Molecular Formula
C8H5F3O
InChI Key
BEOBZEOPTQQELP-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(C=O)C=C1
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Comment
May form precipitate upon standing which does not affect use. Warm gently to redissolve solid.
Free acid (titration)
≤1.5%

Description

4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.

Solubility
Soluble in water. 1.5 g/L at 20°C

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under an inert atmosphere. Incompatible with oxidizing agents, reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text