Thermo Scientific Chemicals

Potassium hydrogen L-tartrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15281.0B
1000 g, Each
Thermo Scientific Chemicals

Potassium hydrogen L-tartrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15281.0B
1000 g, Each
Quantity
Catalog number: A15281.0B
also known as A15281-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
868-14-4
IUPAC Name
potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular Formula
C4H5KO6
InChI Key
KYKNRZGSIGMXFH-ZVGUSBNCSA-M
SMILES
[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
Identification (FTIR)
Conforms
Optical Rotation
32.3 ± 0.4? (C=10 in 1N NaOH)
Appearance (Color)
White to pale cream
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.

Solubility
Soluble in water and dilute mineral acid. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text