Thermo Scientific Chemicals

3-Nitrophthalimide, 98%, Thermo Scientific Chemicals

Catalog number: A15291.14
25 g, Each
Thermo Scientific Chemicals

3-Nitrophthalimide, 98%, Thermo Scientific Chemicals

Catalog number: A15291.14
25 g, Each
Quantity
Catalog number: A15291.14
also known as A15291-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
603-62-3
IUPAC Name
4-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C8H4N2O4
InChI Key
BONIIQYTWOPUQI-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC2=C1C(=O)NC2=O
Appearance (Color)
Pale yellow to yellow
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
215.0-222.0?C

Description

3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text