Metaldehyde, 98%, Thermo Scientific Chemicals
Metaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Metaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number A15292.30
also known as A15292-30
Price (USD)/ Each
55.50
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
55.50
Add to cart
Metaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberA15292.30
Price (USD)/ Each
55.50
-
Add to cart
Chemical Identifiers
CAS108-62-3
IUPAC Name2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
Molecular FormulaC8H16O4
InChI KeyGKKDCARASOJPNG-UHFFFAOYSA-N
SMILESCC1OC(C)OC(C)OC(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only