4'-n-Butylacetophenone is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-n-Butylacetophenone is used as an organic chemical synthesis intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
RUO – Research Use Only
General References:
- Isabelle Kondolff; Henri Doucet; Maurice Santelli. Tetraphosphine/palladium catalysed Suzuki cross-coupling reactions of aryl halides with alkylboronic acids. Tetrahedron. 2004, 60,(17), 3813-3818.
- Phil Ho Lee; Sung Wook Lee; Kooyeon Lee. Pd-catalyzed carbonylative cross-coupling reactions by triorganoindiums: Highly efficient transfer of organic groups attached to indium under atmospheric pressure. Org. Lett. 2003, 5,(7), 1103-1106.