1-(4-Iodophenyl)pyrrole, 97%, Thermo Scientific Chemicals
1-(4-Iodophenyl)pyrrole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Iodophenyl)pyrrole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A15305.06
also known as A15305-06
Price (USD)/ Each
222.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
222.00
Add to cart
1-(4-Iodophenyl)pyrrole, 97%, Thermo Scientific Chemicals
Catalog numberA15305.06
Price (USD)/ Each
222.00
-
Add to cart
Chemical Identifiers
CAS92636-36-7
IUPAC Name1-(4-iodophenyl)-1H-pyrrole
Molecular FormulaC10H8IN
InChI KeyFMURNAZHVQDQQN-UHFFFAOYSA-N
SMILESIC1=CC=C(C=C1)N1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)124.5-133.5?C
Appearance (Color)Cream to brown
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only