Potassium peroxydisulfate, 97%, Thermo Scientific Chemicals
Potassium peroxydisulfate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium peroxydisulfate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A15310.36
also known as A15310-36
Price (USD)/ Each
54.65
Online exclusive
61.00 
Save 6.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
61.00 
Save 6.35 (10%)
Add to cart
Potassium peroxydisulfate, 97%, Thermo Scientific Chemicals
Catalog numberA15310.36
Price (USD)/ Each
54.65
Online exclusive
61.00 
Save 6.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7727-21-1
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or flakes
Assay (Iodometric Titration)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
Potassium peroxydisulfate is used as an oxidizing agent in organic synthesis. It is involved in the Elbs persulfate oxidation of phenols and the Boyland-Sims oxidation of anilines. In solution, it gives radicals and is used to initiate polymerization reactions to prepare styrene-butadiene rubber and polytetrafluoroethylene. It is used as a bleaching agent in various hair bleaches, as an ion exchange agent, a plating agent and a surface treating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium peroxydisulfate is used as an oxidizing agent in organic synthesis. It is involved in the Elbs persulfate oxidation of phenols and the Boyland-Sims oxidation of anilines. In solution, it gives radicals and is used to initiate polymerization reactions to prepare styrene-butadiene rubber and polytetrafluoroethylene. It is used as a bleaching agent in various hair bleaches, as an ion exchange agent, a plating agent and a surface treating agent.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Moisture sensitive. Incompatible with organic materials, strong reducing agents, powdered metals, strong bases, alcohols, phosphorous, anhydrides, halogens and acids.
RUO – Research Use Only

General References:

  1. Useful oxidant: J. Org. Chem., 25, 1901 (1960); examples:
  2. Conversion of phenols to para-diphenols in alkaline solution (Elbs oxidation), and of arylamines to aminophenols (Boyland-Sims oxidation); review: Org. React., 35, 421 (1988). Phase-transfer conversion of acids to peracids in the presence of BTEAC or PEG: Synth. Commun., 18, 2123 (1988). In the presence of Na2HPO4 in acetone - dichloromethane, arylamines give nitro arenes: Tetrahedron Lett., 29, 4501 (1988). In the presence of Ni(II) salts, primary alkylamines can undergo dehydrogenation to nitriles: Bull. Chem. Soc. Jpn., 63, 301 (1990).
  3. See also Ammonium peroxydisulfate, A10533, and Sodium peroxydisulfate, 54100.
  4. Patil, V. V.; Shankarling, G. S. Steric-Hindrance-Induced Regio-and Chemoselective Oxidation of Aromatic Amines. J. Org. Chem. 2015, 80 (16), 7876-7883.
  5. Anirudhan, T. S.; Suchithra, P. S.; Senan, P.; Tharun, A. R. Kinetic and equilibrium profiles of adsorptive recovery of thorium(IV) from aqueous solutions using poly (methacrylic acid) grafted cellulose/bentonite superabsorbent composite. Ind. Eng. Chem. Res. 2012, 51 (13), 4825-4836.