m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals
m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A15313.14
also known as A15313-14
Price (USD)/ Each
87.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
87.50
Add to cart
m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals
Catalog numberA15313.14
Price (USD)/ Each
87.50
-
Add to cart
Chemical Identifiers
CAS620-23-5
IUPAC Name3-methylbenzaldehyde
Molecular FormulaC8H8O
InChI KeyOVWYEQOVUDKZNU-UHFFFAOYSA-N
SMILESCC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Free acid (titration)<2.5%
Identification (FTIR)Conforms
Refractive Index1.5385-1.5435 @ 20?C
View more
m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M.J.Worsey; F.C.Franklin; P.A.Williams. Regulation of the degradative pathway enzymes coded for by the TOL plasmid (pWWO) from Pseudomonas putida mt-2. J. Bacteriol. 1978, 134,(3), 757-764.
  2. K.Furukawa; J.R.Simon; A.M.Chakrabarty. Common induction and regulation of biphenyl, xylene/toluene, and salicylate catabolism in Pseudomonas paucimobilis. J. Bacteriol. 1983, 154,(3), 1356-1362.