Thermo Scientific Chemicals

Nicotinoyl chloride hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: A15317.09
10 g, Each
Thermo Scientific Chemicals

Nicotinoyl chloride hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: A15317.09
10 g, Each
Quantity
Catalog number: A15317.09
also known as A15317-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20260-53-1
IUPAC Name
hydrogen pyridine-3-carbonyl chloride chloride
Molecular Formula
C6H5Cl2NO
InChI Key
MSYBLBLAMDYKKZ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClC(=O)C1=CC=CN=C1
Appearance (Color)
White to cream to gray
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%

Description

Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.

Solubility
It decomposes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text