Thermo Scientific Chemicals

Methyl 2-bromo-5-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A15322.14
25 g, Each
Thermo Scientific Chemicals

Methyl 2-bromo-5-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A15322.14
25 g, Each
Quantity
Catalog number: A15322.14
also known as A15322-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35450-36-3
IUPAC Name
methyl 2-bromo-5-methoxybenzoate
Molecular Formula
C9H9BrO3
InChI Key
VRTQLDFCPNVQNT-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(OC)=CC=C1Br
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5625-1.5665 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Avoid contact with strong oxidizing agents. Store in a cool and dark place. Protect from direct light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text