Thermo Scientific Chemicals

1-Methylnaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: A15327.22
100 g, Each
Thermo Scientific Chemicals

1-Methylnaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: A15327.22
100 g, Each
Quantity
Catalog number: A15327.22
also known as A15327-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-12-0
IUPAC Name
1-methylnaphthalene
Molecular Formula
C11H10
InChI Key
QPUYECUOLPXSFR-UHFFFAOYSA-N
SMILES
CC1=C2C=CC=CC2=CC=C1
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.6120-1.6180 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange to brown
Form
Liquid

Description

1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

Solubility
Miscible with water, alcohol, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text