3,5-Dichlorophenol is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dichlorophenol is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water. Very soluble in ethanol, ethyl ether and petroleum ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
I.M.Arabatzis; S.Antonaraki; T Stergiopoulos; A.Hiskia; E.Papaconstantinou; M.C.Bernard; P.Falaras. Preparation, characterization and photocatalytic activity of nanocrystalline thin film TiO2 catalysts towards 3,5-dichlorophenol degradation. Journal of Photochemistry and Photobiology A: Chemistry. 2002, 149,(1-3), 237-245.
Jacinthe Thibodeau; Annie Gauthier; Marie Duguay; Richard Villemur; Francois Lepine; Pierre Juteau; Rejean Beaudet. Purification, cloning, and sequencing of a 3,5-dichlorophenol reductive dehalogenase from Desulfitobacterium frappieri PCP-1. Appl. Environ. Microbiol. 2004, 70,(8), 4532-4537.