Thermo Scientific Chemicals

4-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A15331.09
10 g, Each
Thermo Scientific Chemicals

4-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A15331.09
10 g, Each
Quantity
Catalog number: A15331.09
also known as A15331-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2284-20-0
IUPAC Name
1-isothiocyanato-4-methoxybenzene
Molecular Formula
C8H7NOS
InChI Key
VRPQCVLBOZOYCG-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)N=C=S
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.6470-1.6510 @ 20?C

Description

4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text