Thermo Scientific Chemicals

Coumarin, 98%, Thermo Scientific Chemicals

Catalog number: A15336.30
250 g, Each
Thermo Scientific Chemicals

Coumarin, 98%, Thermo Scientific Chemicals

Catalog number: A15336.30
250 g, Each
Quantity
Catalog number: A15336.30
also known as A15336-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-64-5
IUPAC Name
2H-chromen-2-one
Molecular Formula
C9H6O2
InChI Key
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
SMILES
O=C1OC2=CC=CC=C2C=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
67.0-73.0?C

Description

Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text