Thermo Scientific Chemicals

Dicyclohexano-18-crown-6, mixture of isomers, 97%, Thermo Scientific Chemicals

Catalog number: A15344.09
10 g, Each
Thermo Scientific Chemicals

Dicyclohexano-18-crown-6, mixture of isomers, 97%, Thermo Scientific Chemicals

Catalog number: A15344.09
10 g, Each
Quantity
Catalog number: A15344.09
also known as A15344-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16069-36-6
IUPAC Name
2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosane
Molecular Formula
C20H36O6
InChI Key
BBGKDYHZQOSNMU-UHFFFAOYNA-N
SMILES
C1CCC2OCCOCCOC3CCCCC3OCCOCCOC2C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

Solubility
Soluble in Toluene. Slightly soluble (13g/L, 26°C) in water.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with strong acids, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text