Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals
Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A15348.14
also known as A15348-14
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals
Catalog numberA15348.14
Price (USD)/ Each
102.00
-
Add to cart
Chemical Identifiers
CAS18595-18-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)110.0-119.0?C
Appearance (Color)White to cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only