Thermo Scientific Chemicals

Dimethyl oxalate, 99%, Thermo Scientific Chemicals

Catalog number: A15359.36
500 g, Each
Thermo Scientific Chemicals

Dimethyl oxalate, 99%, Thermo Scientific Chemicals

Catalog number: A15359.36
500 g, Each
Quantity
Catalog number: A15359.36
also known as A15359-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
553-90-2
IUPAC Name
dimethyl oxalate
Molecular Formula
C4H6O4
InChI Key
LOMVENUNSWAXEN-UHFFFAOYSA-N
SMILES
COC(=O)C(=O)OC
Appearance (Color)
White
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Water Content (Karl Fischer Titration)
≤0.75%

Description

Dimethyl oxalate is used as an alternate fuel in the direct oxidation fuel cell. It is involved in the catalytic hydrogenation using Cu/SiO2 to prepare ethylene glycol. It is also used in the selective gas-phase hydrogenation catalyzed by Ag/SiO2 to get the corresponding alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl oxalate is used as an alternate fuel in the direct oxidation fuel cell. It is involved in the catalytic hydrogenation using Cu/SiO2 to prepare ethylene glycol. It is also used in the selective gas-phase hydrogenation catalyzed by Ag/SiO2 to get the corresponding alcohol.

Solubility
Soluble in ethanol. Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text