4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
50 g
250 g
Catalog number A15364.0B
also known as A15364-0B
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
Add to cart
4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberA15364.0B
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS123-11-5
IUPAC Name4-methoxybenzaldehyde
Molecular FormulaC8H8O2
InChI KeyZRSNZINYAWTAHE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Free acid (titration)≤1.0%
Refractive Index1.5710-1.5750 @ 20?C
View more
4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.

Solubility
Miscible with acetone, alcohol, ether, chloroform and benzene. Immiscible with water.

Notes
Air and Light Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

General References:

  1. Diols, under acid catalysis, can be protected as 4-methoxybenzylidene cyclic acetals, hydolyzed ca 10x faster by acid than the benzylidene acetal: J. Am. Chem. Soc., 84, 430 (1962). Cleavage with Ce(IV): J. Chem. Soc., Chem. Commun., 2010 (1984), hydrogenolysis: Tetrahedron Lett., 32, 6155 (1991), or DibalH: Tetrahedron Lett., 28, 3835 (1987); Tetrahedron: Asymmetry, 6, 2319 (1995) have also been reported. See also 4-Methoxybenzaldehyde dimethyl acetal, A15793.
  2. Brullo, C.; Massa, M.; Rocca, M.; Rotolo, C.; Guariento, S.; Rivera, D.; Ricciarelli, R.; Fedele, E.; Fossa, P.; Bruno, O. Synthesis, Biological Evaluation, and Molecular Modeling of New 3-(Cyclopentyloxy)-4-methoxybenzaldehyde O-(2-(2,6-Dimethylmorpholino)-2-oxoethyl) Oxime (GEBR-7b) Related Phosphodiesterase 4D (PDE4D) Inhibitors. J. Med. Chem. 2014, 57 (16), 7061-7072.
  3. Denizalti, S.; Turkmen, H.; Cetinkaya, B. Chelating alkoxy NHC-Rh(I) complexes and their applications in the arylation of aldehydes. Tet. Lett. 2014, 55 (30), 4129-4132.