Thermo Scientific Chemicals

4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15364.18
50 g, Each
Thermo Scientific Chemicals

4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15364.18
50 g, Each
Quantity
Catalog number: A15364.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-11-5
IUPAC Name
4-methoxybenzaldehyde
Molecular Formula
C8H8O2
InChI Key
ZRSNZINYAWTAHE-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=O)C=C1
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Free acid (titration)
≤1.0%
Refractive Index
1.5710-1.5750 @ 20?C

Description

4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.

Solubility
Miscible with acetone, alcohol, ether, chloroform and benzene. Immiscible with water.

Notes
Air and Light Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text