Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.
Solubility
Soluble in NH{4}OH
Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
RUO – Research Use Only
Jianfang Cao; Shuxia Liu; Ruige Cao; Linhua Xie; Yuanhang Ren; Chaoying Gao and Lin Xu. Organic-inorganic hybrids assembled by bis(undecatungstophosphate) lanthanates and dinuclear copper(II)-oxalate complexes.Dalton Trans.2008, 115-120.
Kenneth T. McGregor; Zoltan G. Soos. Anisotropic exchange and temperature-dependent electron paramagnetic resonance line width in one-dimensional copper(II) complexes. 2. Magnetic properties of copper(II) oxalate-1/3-water.Inorg. Chem.1976, 15 (9), 2159-2165.