Thermo Scientific Chemicals

2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog number: A15370.06
5 g, Each
Thermo Scientific Chemicals

2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog number: A15370.06
5 g, Each
Quantity
Catalog number: A15370.06
also known as A15370-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6482-24-2
IUPAC Name
1-bromo-2-methoxyethane
Molecular Formula
C3H7BrO
InChI Key
YZUPZGFPHUVJKC-UHFFFAOYSA-N
SMILES
COCCBr
Assay (GC)
≥95.0%
Refractive Index
1.4450-1.4510 @ 20°C
Appearance (Color)
Clear colourless to yellow
Form
Liquid

Description

2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

Solubility
Soluble in water.14,4 g/L at 25°C

Notes
Protect from heat. Incompatible with heat, strong bases and oxidizing agents. Store at 2-8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text