Thermo Scientific Chemicals

4-Fluoro-3-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A15371.14
25 g, Each
Thermo Scientific Chemicals

4-Fluoro-3-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A15371.14
25 g, Each
Quantity
Catalog number: A15371.14
also known as A15371-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
364-76-1
IUPAC Name
4-fluoro-3-nitroaniline
Molecular Formula
C6H5FN2O2
InChI Key
LLIOADBCFIXIEU-UHFFFAOYSA-N
SMILES
NC1=CC=C(F)C(=C1)[N+]([O-])=O
Assay (GC)
≥97.5%
Appearance (Color)
Yellow to yellow-brown
Identification (FTIR)
Conforms
Form
Powder
Melting Point
94.5-99.5?C

Description

4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Incompatible with Oxidizing agents, Acids, Acid chlorides, Acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text