Thermo Scientific Chemicals

Tri(m-tolyl)phosphine, 98+%, Thermo Scientific Chemicals

Catalog number: A15372.03
1 g, Each
Thermo Scientific Chemicals

Tri(m-tolyl)phosphine, 98+%, Thermo Scientific Chemicals

Catalog number: A15372.03
1 g, Each
Quantity
Catalog number: A15372.03
also known as A15372-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6224-63-1
IUPAC Name
tris(3-methylphenyl)phosphane
Molecular Formula
C21H21P
InChI Key
LFNXCUNDYSYVJY-UHFFFAOYSA-N
SMILES
CC1=CC(=CC=C1)P(C1=CC(C)=CC=C1)C1=CC=CC(C)=C1
Assay (GC)
≥98.0%
Form
Powder
Melting Point (clear melt)
98.0-105.0?C
Appearance (Color)
White

Description

Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text