Thermo Scientific Chemicals

2-Chloroethanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A15377.18
50 g, Each
Thermo Scientific Chemicals

2-Chloroethanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A15377.18
50 g, Each
Quantity
Catalog number: A15377.18
also known as A15377-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1622-32-8
IUPAC Name
2-chloroethane-1-sulfonyl chloride
Molecular Formula
C2H4Cl2O2S
InChI Key
VHCSBTPOPKFYIU-UHFFFAOYSA-N
SMILES
ClCCS(Cl)(=O)=O
Form
Liquid
Assay (Titration ex Chloride)
≥97.5 to ≤102.5% (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Appearance (Color)
Clear colorless
Assay from Suppliers CofA
≥97.5% (U.S. specification)

Description

2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text