Thermo Scientific Chemicals

3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15388.14
25 g, Each
Thermo Scientific Chemicals

3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15388.14
25 g, Each
Quantity
Catalog number: A15388.14
also known as A15388-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5973-71-7
IUPAC Name
3,4-dimethylbenzaldehyde
Molecular Formula
C9H10O
InChI Key
POQJHLBMLVTHAU-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=O)C=C1C
Refractive Index
1.5480-1.5530 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Form
Liquid

Description

3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

Solubility
Soluble in ethanol, ether, toluene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text