Thermo Scientific Chemicals

2-Chloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: A15394.22
100 g, Each
Thermo Scientific Chemicals

2-Chloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: A15394.22
100 g, Each
Quantity
Catalog number: A15394.22
also known as A15394-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1722-12-9
IUPAC Name
2-chloropyrimidine
Molecular Formula
C4H3ClN2
InChI Key
UNCQVRBWJWWJBF-UHFFFAOYSA-N
SMILES
ClC1=NC=CC=N1
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Appearance (Color)
White to pale brown or pale yellow to yellow to pale orange
Identification (FTIR)
Conforms
Melting Point (clear melt)
62-67?C

Description

2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4?-(1,1?-(5-(2-methoxyphenoxy)-[2,2?-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4′-(1,1′-(5-(2-methoxyphenoxy)-[2,2′-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

Solubility
Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text