Thermo Scientific Chemicals

2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A15397.36
500 g, Each
Thermo Scientific Chemicals

2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A15397.36
500 g, Each
Quantity
Catalog number: A15397.36
also known as A15397-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-05-1
IUPAC Name
2,6-dibromopyridine
Molecular Formula
C5H3Br2N
InChI Key
FEYDZHNIIMENOB-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(Br)=N1
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to cream to pale brown to gray
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.

Solubility
Soluble in hot methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text