1,8-Octanediol, 98+%, Thermo Scientific Chemicals
1,8-Octanediol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,8-Octanediol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15402.14
also known as A15402-14
Price (USD)/ Each
50.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
50.80
Add to cart
1,8-Octanediol, 98+%, Thermo Scientific Chemicals
Catalog numberA15402.14
Price (USD)/ Each
50.80
-
Add to cart
Chemical Identifiers
CAS629-41-4
IUPAC Nameoctane-1,8-diol
Molecular FormulaC8H18O2
InChI KeyOEIJHBUUFURJLI-UHFFFAOYSA-N
SMILESOCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystalline powder or crystals or flakes or pellets
Melting Point (clear melt)58-63?C
Solution Test5% w/v in methanol: clear
Assay (GC)≥98.0%
View more
1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents, reducing agents, acid chlorides acid, anhydrides and chloroformates.
RUO – Research Use Only

General References:

  1. Qian, F.; Luo, Q.; Yang, R.; Zhu, Z.; Chen, H.; Yan, X. The littoral red alga Pyropia haitanensisuses rapid accumulation of floridoside as the desiccation acclimation strategy. J. Appl. Phycol. 2015, 27 (1), 621-632.
  2. Zeimaran, E.; Pourshahrestani, S.; Pingguan-Murphy, B.; Kadri, N. A.; Rothan, H. A.; Yusof, R.; Towler, M. R.; Djordjevic, I. Fabrication and characterization of poly(octanediol citrate)/gallium-containing bioglass microcomposite scaffolds. J. Mater. Sci. 2015, 50 (5), 2189-2201.